Learn More
3M™ Microbial Luminescence System Microwell Strip 3008, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S27262 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
320/Cs.
Catalog No. Quantity
S27262 320/Cs.
1 options
Catalog No. S27262 Supplier 3M™ Supplier No. 3008
Add to Cart
Add to Cart
For use with 3M™ Microbial Luminescence System (MLS)
  • Plastic strips of 12 wells
  • Snap to desired number of wells
  • Use with 3M™ MLS Microwell Strip Holder #3009
  • Used for reagent checks and low-volume UHT testing
Specifications
Product Type Plastic Strips of 12 Wells
For Use With (Equipment) 3M™ MLS Microwell Strip Holder #3009 and 3M™ Microbial Luminescence System (MLS)
For Use With (Application) Reagent checks and UHT testing
Quantity 320/Cs.