Learn More
3M™ Microbial Luminescence System MLSII, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S27260 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Detects:
ATP
Catalog No. Detects
S27260 ATP
1 options
Catalog No. S27260 Supplier 3M™ Supplier No. MLSII
Add to Cart
Add to Cart
Request a Quote
Uses ATP bioluminescence in microwell based system to detect microbial ATP in Ultra Heat Treated (UTH) and Extended Shelf Life (ESL) dairy and dairy end products
  • Microplate, 5 injector-dispenser luminometer designed for microbial detection
  • Results available approximately 48 hours sooner than traditional methods
  • Reduces stock holding requirements and improves responsiveness to customers
  • Time saved improves cash flow
  • Allows quality performance to be tracked over time
Specifications
Product Type Microbial Luminescence System
Detects ATP
For Use With (Application) Screening microbial ATP in UTH and ESL dairy and dairy end products