Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Microbial Luminescence System Ultra High Temperature Beverage Screen Kit BEV600, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S27267 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
600 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27267 600 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27267 Supplier 3M™ Supplier No. BEV600
Add to Cart
Add to Cart

Use with 3M™ Microbial Luminescence System (MLS) for testing Ultra high temperature (UHT) and ESL beverages

  • Eliminates second incubation period needed with traditional methods
  • Reagents for 600 tests

Specifications

Format Kit
Product Type Food Testing Media
For Use With (Application) 3M™ Microbial Luminescence System
Quantity 600 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.