Learn More
Sigma Aldrich Fine Chemicals Biosciences 96 Well Low temp PCR rack
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS120538
Encompass
The PCR storage rack is versatile it can be used on a bench top or for freezer storage. The compact footprint is stackable which helps save valuable space in the lab. Ergonomic textured finger grips provide a secure hold reducing the chance of dropping the PCR rack and makes handling and transportation easy especially when using gloves. The lid has a side opening that enables faster cooling of samples to ensure integrity and is made from translucent polycarbonate for easy sample and well identification.The PCR rack base is made from robust ABS construction that helps keep samples safe so you dont have to spend extra money and time on expensive retesting. A location ID of numbers and letters allows for organized sample placement which results in easy and quick sample retrieval. The PCR rack has a closed base design that helps samples integrity by staying cold longer.The PCR storage rack is versatile it can be used on a bench top or for freezer storage. The compact footprint is stack
Catalog No. 50-290-1711
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.