Learn More
Microbiologics™ Cepheid Xpert GBS QC Set
SDP
Catalog No. 23002133 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
4/Pk.
Catalog No. Quantity
23-002-133 4/Pk.
1 options
Catalog No. 23-002-133 Supplier Microbiologics™ Supplier No. 8164
Add to Cart
Add to Cart
Includes
E3 Streptococcus species (Group B, Ib), ATCC™ 12401™†, E5 Streptococcus species (Group B, Ib), ATCC™ 12401™†, E7 Streptococcus species (Group B, Ib), ATCC™ 12401™†, Neg Control Lactobacillus acidophilus, ATCC ™4356™†
Microorganism sets for culture purposes and QC applications
  • Commercial sets contain the manufacturer recommended quality control strains for bacterial identification systems
  • Ordering microorganisms in sets simplifies the ordering process
  • One convenient packaging option
  • Four KWIK-STIK™ units; each KWIK-STIK device features a single microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • Each set contains a low, medium and high level of the same microorganism and one negative control
  • For the use with Cepheid™ systems only
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
Product Type KWIK-STIK
Quantity 4/Pk.