Learn More
Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features single strain of Enterobacter cloacaei in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
- For added convenience, peel-off identification label is included for easy documentation
- Available in pack of 2 or 6
- Reference stock culture option; lyophilized pellet containing single strain of Enterobacter cloacae
- Easy to use; re-hydrate pellet and inoculate
- Available in re-sealable vial containing 6 pellets
- NCTC 13406
- Control for positive test of AmpC detection
Specifications
Specifications
| ATCC® | NCTC 13406 |
| Biosafety Level | 2 |
| Format | LYFO-DISK |
| Product Type | Quality Control Organism |
| Quantity | 6/Pk. |
| Species | Enterobacter cloacae |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
