Learn More
Microbiologics™ Epower™ Bacillus cereus ATCC™ 10876™†
SDP
Catalog No. 12111236 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
12-111-236 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 12-111-236 Supplier Microbiologics™ Supplier No. 0998E3CRM
Add to Cart
Add to Cart
This item is not returnable. View return policy
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 10876™†
Specifications
ATCC® 10876™*
Biosafety Level 1
Form Lyophilized Pellet
Quantity 10/Pk.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.