Learn More
Microbiologics™ Epower™ Bacillus subtilis subsp. spizizenii ATCC™ 6633™†
SDP
Catalog No. 12111239 Shop All Microbiologics Products
Change view
Click to view available options
Mean Assay Value:
1.0-9.9 + 3 CFU/pellet
1.0-9.9 + 4 CFU/pellet
1.0-9.9 + 6 CFU/pellet
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mean Assay Value
12-111-239 1.0-9.9 + 4 CFU/pellet
12-111-238 1.0-9.9 + 3 CFU/pellet
12-111-240 1.0-9.9 + 6 CFU/pellet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 12-111-239 Supplier Microbiologics™ Supplier No. 0486E4CRM
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 6633™†
Specifications
ATCC® 6633™*
Biosafety Level 1
Form Lyophilized Pellet
Mean Assay Value 1.0-9.9 + 4 CFU/pellet
Quantity 10/Pk.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures