Learn More
Microbiologics™ Epower™ Klebsiella aerogenes ATCC™ 13048™
SDP
Catalog No. 12111248 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
12-111-248 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 12-111-248 Supplier Microbiologics™ Supplier No. 0306E3CRM
Add to Cart
Add to Cart
This item is not returnable. View return policy
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 13048™†
Specifications
ATCC® 13048
Biosafety Level 1
Form Lyophilized Pellet
For Use With (Application) For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
Mean Assay Value 1.0-9.9 + 3 CFU/pellet
Quantity 10/Pk.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.