Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Epower Escherichia coli serotype O103:H11 CDC 06-3008 (STEC)
SDP

Catalog No. 01101E3 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-101-E3 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-101-E3 Supplier MicroBioLogics Supplier No. 01101E3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Assay Range 1.0-9.9E+03CFU/pellet
Biosafety Level 2
Product Type Escherichia coli
Quantity 10/Pk.
Species Escherichia coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.