Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Epower™ Pseudomonas aeruginosa, ATCC™ 9027™†
SDP

Catalog No. 23004531 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Pseudomonas aeruginosa
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-004-531 Pseudomonas aeruginosa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-004-531 Supplier Microbiologics™ Supplier No. 0484E7
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Product warranty and peel-off certificate of assay

Lyophilized pellets add convenience to special projects and internal proficiency plans.

  • Biosafety level: 2
  • Mean Assay Value: 1.0-9.9E+07 CFU per pellet

Order Info

Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet

Specifications

ATCC® 9027
Biosafety Level 2
Form Lyophilized Pellet
Product Type QC Microorganism
For Use With (Application) Lyophilized pellets add convenience to special projects and internal proficiency plans.
Includes Product warranty and peel-off certificate of assay
Mean Assay Value 0-9.9E+07 CFU per pellet
Packaging Type Vial
Quantity 10/Pk.
Species Pseudomonas aeruginosa

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.