Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Epower™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™†
SDP

Catalog No. 12111285 Shop All Microbiologics Products
Change view
Click to view available options
Mean Assay Value:
1-9.9 + 3 CFU/pellet
1.0-9.9E + 04 CFU/pellet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mean Assay Value
12-111-285 1-9.9 + 3 CFU/pellet
12-111-286 1.0-9.9E + 04 CFU/pellet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-111-285 Supplier Microbiologics™ Supplier No. 0363E3CRM
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy

For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination

Epower

  • Lyophilized microorganism preparations
  • Biosafety Level: 2
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 14028™†

 

Specifications

ATCC® 14028
Biosafety Level 2
Form Lyophilized Pellet
For Use With (Application) For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination.
Mean Assay Value 1-9.9 + 3 CFU/pellet
Quantity 10/Pk.
Species Salmonella enterica subsp. enterica serovar typhimurium

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.