Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Epower™ Staphylococcus aureus ATCC™ 700699™†
SDP

Catalog No. 23015305 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Staphylococcus aureus subsp. aureus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-015-305 Staphylococcus aureus subsp. aureus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-015-305 Supplier Microbiologics™ Supplier No. 0158E4
Add to Cart
Add to Cart
This item is not returnable. View return policy

For culture purposes and QC applications

Epower™

  • 10 pellets of a microorganism manufactured with a predetermined quantitative assay
  • Can be used to perform many essential quality control functions including microbial detection and enumeration, equipment calibration, method validation, bioburden determination, and antibacterial effectiveness and lethality testing
  • Versatile lyophilized microorganism preparations available in concentrations ranging from 102 to 108 CFU per pellet
  • Can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • For added convenience, a peel-off Certificate of Assay is included for quality control documentation
  • Packaged in a resealable vial containing 10 pellets of a single quantitative QC microorganism strain
  • ATCC™ 700699™†
  • Methicillin resistant
  • Biosafety level 2
  • Mean Assay Value: 1.0-9.9E+04 CFU per pellet

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Specifications

ATCC® 700699™*
Biosafety Level 2
Form Lyophilized Pellet
Quantity 10/Pk.
Species Staphylococcus aureus subsp. aureus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.