Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Escherichia coli ATCC™ BAA-2469™†
SDP

Catalog No. 23017183 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2/Pk.
6/Pk.
Format:
KWIK-STIK
LYFO-DISK
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
23-017-183 6/Pk. KWIK-STIK
23-017-185 2/Pk. KWIK-STIK
23-017-184 6/Pk. LYFO-DISK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-017-183 Supplier Microbiologics™ Supplier No. 01113K
Add to Cart
Add to Cart

For culture purposes and QC applications

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features single strain of Escherichia coli in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
  • For added convenience, peel-off identification label is included for easy documentation
  • Available in pack of 2 or 6
LYFO DISK™
  • Reference stock culture option; lyophilized pellet containing single strain of Escherichia coli
  • Easy to use; re-hydrate pellet and inoculate
  • Available in re-sealable vial containing 6 pellets
  • ATCC™ BAA-2469™†
  • New Delhi metallo-beta-lactamase (NDM-1) positive; blaKPC negative by PCR; blaNDM positive by PCR; Carbepenem-resistant (Imipenem and Ertapenem)

Specifications

ATCC® ATCC BAA-2469
Format KWIK-STIK
Product Type Quality Control Organism
Quantity 6/Pk.
Species Escherichia coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.