Learn More
Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features single strain of Escherichia coli in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
- For added convenience, peel-off identification label is included for easy documentation
- Available in pack of 2 or 6
- Reference stock culture option; lyophilized pellet containing single strain of Escherichia coli
- Easy to use; re-hydrate pellet and inoculate
- Available in re-sealable vial containing 6 pellets
- ATCC™ BAA-2469™†
- New Delhi metallo-beta-lactamase (NDM-1) positive; blaKPC negative by PCR; blaNDM positive by PCR; Carbepenem-resistant (Imipenem and Ertapenem)
Specifications
Specifications
| ATCC® | ATCC BAA-2469 |
| Format | LYFO-DISK |
| Product Type | Quality Control Organism |
| Quantity | 6/Pk. |
| Species | Escherichia coli |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
