Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ EZ-CFU™ Aspergillus brasiliensis ATCC™ 16404™
SDP

Catalog No. 23001264 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Aspergillus brasiliensis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-001-264 Aspergillus brasiliensis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-264 Supplier Microbiologics™ Supplier No. 0392C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Product warranty and peel-off certificate of assay

Assayed microorganism preparation for USP quantitative microorganism challenges

Lyophilized microorganism preparations delivering the desired 10 to 100CFU/0.1mL of challenge material (following hydration and dilution of 1.0mL of hydrated suspension in 9.0mL of pH 7.2 phosphate buffer).

  • Traceable to reference cultures

Order Info

Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

Specifications

ATCC® 16404
Format EZ-CFU
Product Type EZ-CFU
For Use With (Application) Assayed microorganism preparations for USP quantitative microorganism challenges.
Includes Product warranty and peel-off certificate of assay
Packaging Type Vial
Species Aspergillus brasiliensis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.