Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ EZ-CFU™ One-Step Microorganism Preparations
SDP

Catalog No. 23004841 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Aspergillus brasiliensis
Bacillus cereus
Bacillus subtilis subsp. spizizenii
Candida albicans
Clostridium sporogenes
Enterobacter aerogenes
Escherichia coli
Geobacillus stearothermophilus
Micrococcus luteus
Pseudomonas aeruginosa
Salmonella enterica subsp. enterica serovar abony
Salmonella enterica subsp. enterica serovar typhimurium
Staphylococcus aureus subsp. aureus
Staphylococcus epidermidis
Streptococcus pyogenes
ATCC®:
10231
10876
11437
12228
13048
14028
16404™*
19404
19615
25923
27853
6538
6538P
6633
7953
8739
9027
ATCC 4698
NCTC 6017
Format:
EZ-CFU
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species ATCC® Format
23-004-841 Aspergillus brasiliensis 16404™* EZ-CFU
23-001-898 Bacillus subtilis subsp. spizizenii 6633 EZ-CFU
23-001-899 Candida albicans 10231 EZ-CFU
23-001-902 Escherichia coli 8739 EZ-CFU
23-001-901 Clostridium sporogenes 19404 EZ-CFU
23-001-900 Clostridium sporogenes 11437 EZ-CFU
23-001-904 Staphylococcus aureus subsp. aureus 6538 EZ-CFU
23-001-903 Pseudomonas aeruginosa 9027 EZ-CFU
23-003-827 Micrococcus luteus ATCC 4698 EZ-CFU
23-003-835 Salmonella enterica subsp. enterica serovar abony NCTC 6017 EZ-CFU
23-003-836 Salmonella enterica subsp. enterica serovar typhimurium 14028 EZ-CFU
23-003-838 Staphylococcus epidermidis 12228 EZ-CFU
23-003-843 Streptococcus pyogenes 19615 EZ-CFU
23-003-844 Staphylococcus aureus subsp. aureus 25923 EZ-CFU
23-003-848 Pseudomonas aeruginosa 27853 EZ-CFU
23-003-847 Geobacillus stearothermophilus 7953 EZ-CFU
23-004-524 Staphylococcus aureus subsp. aureus 6538P EZ-CFU
23-004-525 Bacillus cereus 10876 EZ-CFU
23-004-838 Enterobacter aerogenes 13048 EZ-CFU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Catalog No. 23-004-841 Supplier Microbiologics™ Supplier No. 0392Z
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Product warranty and a peel-off Certificate of Assay

Lyophilized microorganism preparations deliver less than 100CFU per 0.1mL of hydrated suspension—no dilutions are required

EZ-CFU One-Step preparations are designed to provide the required microorganism challenge concentrations for tests such as Growth Promotion Tests, Microbial Limits Tests and Microbial Enumeration Tests. These ready-to-use, dependable, and cost-effective enumerated microorganism preparations allow laboratories to conveniently and confidently conquer a variety of mandated Pharmacopoeial challenges.

  • Up to 19 tests per 2mL aliquot or 190 total tests per kit can be completed
  • Produces a durable, non-foaming suspension that is ready to be added to the challenge material immediately following the hydration step (30 minutes)
  • For added convenience, a peel-off Certificate of Assay is included for quality control documentation
  • Traceable to reference cultures

Order Info

Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

Specifications

ATCC® 16404™*
Format EZ-CFU
Product Type EZ-CFU
Includes Product warranty and peel-off certificate of assay
Packaging Type Vial
Species Aspergillus brasiliensis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.