Learn More
Microbiologics™ EZ-FPC™ Arthrobacter psychrolactophilus ATCC™ 700733™†
SDP
Catalog No. 23016106 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Arthrobacter psychrolactophilus
Catalog No. Species
23-016-106 Arthrobacter psychrolactophilus
1 options
This item is not returnable. View return policy
Catalog No. 23-016-106 Supplier Microbiologics™ Supplier No. 0260K
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quantitative samples for daily process control
EZ-FPC™
  • Food process controls provide the dependability and reliability food testing laboratories need to meet the requirements of quality assurance programs and ensure the health of consumers
  • Lyophilized Arthrobacter psychrolactophilus preparations for daily process control for quantitative detection methods
  • Quantitative preparations provide a guaranteed concentration of 103 CFU per pellet
  • For added convenience, a peel-off Certificate of Assay is included for quality control documentation
  • Packaged in a vial of 10 pellets containing Arthrobacter psychrolactophilus
  • Quantitative Preparations at a concentration of 1.0-9.9E+03 CFU per pellet
  • ATCCS™ 700733™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® ATCC 700733
Product Type KWIK-STIK
Quantity 6/Pk.
Species Arthrobacter psychrolactophilus