Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ EZ-PEC™ Staphylococcus aureus NCIMB Number 9518
SDP

Catalog No. 23015326 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Staphylococcus aureus subsp. aureus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-015-326 Staphylococcus aureus subsp. aureus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-015-326 Supplier Microbiologics™ Supplier No. 0579PEC

EZ-PEC packaging

  • 2 vials [10 pellets each]of a microorganism delivering a final concentration of 100,000 to 1,000,000 CFU/mL
  • 10 vials [2.0mL each] of hydrating fluid
  • Biosafety level 2
  • NCIMB Number 9518

Specifications

ATCC® NCIMB 9518™*
Biosafety Level 2
Format EZ-PEC
Packaging Type Vial
Quantity 2 Vials of 10 Pellets
Species Staphylococcus aureus subsp. aureus

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.