Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Bacillus cereus ATCC™ 11778™†
SDP

Catalog No. 23003849 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Bacillus cereus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-003-849 Bacillus cereus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-003-849 Supplier Microbiologics™ Supplier No. 0256SPR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enumerated lyophilized microorganism preparations are used as daily process controls for qualitative and quantitative spoilage detection methods

EZ-SPORE™
  • Provides a guaranteed concentration of 104CFU per pellet
  • Quantitative daily process controls
  • Maintenance free
  • Traceable to reference culture
  • For added convenience, a peel-off Certificate of Assay is included for quality control documentation
  • Packaged in a vial of 10 pellets of a single lyophilized strain
  • Not for use as a verification for steam autoclave sterilization

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures

Specifications

ATCC® 11778
Biosafety Level 1
Format EZ-SPORE
Product Type EZ-SPORE
For Use With (Application) Enumerated lyophilized microorganism preparations are used as daily process controls for qualitative and quantitative spoilage detection methods.
Includes Product warranty and peel-off certificate of assay
Packaging Type Vial
Quantity 10/Pk.
Species Bacillus cereus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.