Learn More
Microbiologics™ Corynebacterium urealyticum ATCC™ 43044™†
SDP
Catalog No. 23016450 Shop All Microbiologics Products
Change view
Click to view available options
Product Type:
KWIK-STIK
Lyfo-Disk QC Control
Quantity:
2/Pk.
6/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
23-016-450 Lyfo-Disk QC Control 6/Pk.
23-002-108 KWIK-STIK 2/Pk.
23-016-014 KWIK-STIK 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-016-450 Supplier Microbiologics™ Supplier No. 01039L
Add to Cart
Add to Cart
This item is not returnable. View return policy
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Corynebacterium urealyticum strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Corynebacterium urealyticum strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 43044™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® 43044
Biosafety Level 2
Product Type Lyfo-Disk QC Control
Quantity 6/Pk.
Species Corynebacterium urealyticum