Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Klebsiella pneumoniae subsp. pneumoniae ATCC™ 4352™†
SDP

Catalog No. 2300173 Shop All Microbiologics Products
Change view
Click to view available options
Product Type:
Lyfo-Disk QC Control
Quality Control Organism
Quantity:
2/Pk.
5/Pk.
6/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
23-001-73 Quality Control Organism 2/Pk.
23-016-301 Quality Control Organism 6/Pk.
23-016-776 Lyfo-Disk QC Control 6/Pk.
23-017-158 Quality Control Organism 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-001-73 Supplier Microbiologics™ Supplier No. 0683P
Add to Cart
Add to Cart

For culture purposes and QC applications

  • Biosafety Level 2
  • Methicillin resistant
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features single strain of Klebsiella pneumoniae subsp. pneumoniae in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
  • For added convenience, peel-off identification label is included for easy documentation
  • Available in packs of 2 and 6
KWIK-STIK™ Plus
  • Five self-contained KWIK-STIK units with microorganism limited to two passages from reference culture
LYFO DISK™
  • Most economical reference stock culture option; lyophilized pellet containing single strain of Klebsiella pneumoniae subsp. pneumoniae
  • Easy to use, simply re-hydrate pellet and inoculate
  • Available in re-sealable vial containing 6 or 10 pellets
  • ATCC™ 4352™†

Specifications

ATCC® 4352
Biosafety Level 2
Certifications/Compliance CE, online certificate of analysis
Content And Storage 2°C to 8°C
Form Lyophilized Pellet
Media Type Kwik-stik™
Product Type Quality Control Organism
For Use With (Application) Culture purpose and QC application
Quantity 2/Pk.
Species Klebsiella pneumoniae

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.