Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Listeria monocytogenes Cornell University SLR2249
SDP

Catalog No. 23016102 Shop All Microbiologics Products
Change view
Click to view available options
Format:
EZ-FPC
KWIK-STIK
LYFO DISK
Quantity:
10/Pk.
6/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Quantity
23-016-102 KWIK-STIK 6/Pk.
23-016-543 LYFO DISK 6/Pk.
23-004-836 EZ-FPC 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-016-102 Supplier Microbiologics™ Supplier No. 0254K
Add to Cart
Add to Cart
This item is not returnable. View return policy

For culture purposes and QC applications

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single Listeria monocytogenes strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Listeria monocytogenes strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
EZ-FPC™
  • Food process controls provide the dependability and reliability food testing laboratories need to meet the requirements of quality assurance programs and ensure the health of consumers
  • Qualitative; 100 to 1,000 CFU per pellet
  • Act A removed
  • Packaged in a vial of 10 pellets containing

Specifications

ATCC® ATCC SLR2249
Biosafety Level 2
Certifications/Compliance CE, online certificate of analysis
Content And Storage 2°C to 8°C
Form Lyophilized Pellet
Format KWIK-STIK
Product Type Quality Control Organism
For Use With (Application) Culture purpose and QC application
Quantity 6/Pk.
Species Listeria monocytogenes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.