Learn More
Microbiologics™ Microbacterium liquefaciens ATCC™ BAA-1819™†
SDP
Catalog No. 23002117 Shop All Microbiologics Products
Change view
Click to view available options
Product Type:
KWIK-STIK
Quantity:
2/Pk.
6/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
23-002-117 KWIK-STIK 2/Pk.
23-016-017 KWIK-STIK 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-002-117 Supplier Microbiologics™ Supplier No. 01042P
Add to Cart
Add to Cart
This item is not returnable. View return policy
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Microbacterium liquefaciens strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Microbacterium liquefaciens strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ BAA-1819™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® BAA-1819™*
Biosafety Level 1
Product Type KWIK-STIK
Includes Peel-off identification label
Packaging Type Re-sealable Vial
Quantity 2/Pk.
Species Microbacterium liquefaciens