Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Zygosaccharomyces rouxii NCYC 381
SDP

Catalog No. 23016103 Shop All Microbiologics Products
Change view
Click to view available options
Format:
KWIK-STIK
LYFO-DISK
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-016-103 KWIK-STIK
23-016-544 LYFO-DISK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-016-103 Supplier Microbiologics™ Supplier No. 0255K
Add to Cart
Add to Cart

For culture purposes and QC applications

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single Zygosaccharomyces rouxii stain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 2 or 6
KWIK-STIK Plus™
  • Available in pack of 1
  • LYFO DISK™
    • The most economic2al reference stock culture option; lyophilized pellet containing a single Zygosaccharomyces rouxii stain
    • Easy to use; simply re-hydrate the pellet and inoculate
    • Available in a re-sealable vial containing 6 pellets
    • NCYC 381

    Specifications

    ATCC® NCYC 381
    Biosafety Level 1
    Certifications/Compliance CE, online certificate of analysis
    Content And Storage 2°C to 8°C
    Form Lyophilized Pellet
    Format KWIK-STIK
    Product Type Quality Control Organism
    For Use With (Application) Culture purpose and QC application
    Quantity 6/Pk.
    Species Zygosaccharomyces rouxii
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.