Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Lab-Elite™ Escherichia coli ATCC™ 11229™†
SDP

Catalog No. 23002059 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Escherichia coli
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-002-059 Escherichia coli
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-002-059 Supplier Microbiologics™ Supplier No. 0681CRM
Add to Cart
Add to Cart

For validation and verification of processes and instruments

Lab Elite

  • Delivered in an easy-to-use KWIK-STIK™ format
  • Traceable to reference culture
  • Highly characterized Escherichia coli
  • For use in ISO 17025 accredited labs and supported by the highest accreditations in the industry
  • ATCC™ 11229™†

Specifications

ATCC® 11229™*
Biosafety Level 1
Format KWIK-STIK
For Use With (Application) For validation and verification of processes and instruments.
Includes Lyophilized microbe pellet, reservoir of hydrating fluid, inoculating swab (KWIK-STIK format)
Packaging Type Single
Species Escherichia coli

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.