Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Lab-Elite™ Escherichia coli ATCC™ 43888
SDP

Catalog No. 12111360 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Escherichia coli
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
12-111-360 Escherichia coli
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-111-360 Supplier Microbiologics™ Supplier No. 0795CRM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Biosafety Level 2
Format KWIK-STIK™
For Use With (Application) For validation and verification of processes and instruments.
Includes Lyophilized microbe pellet, reservoir of hydrating fluid, inoculating swab (KWIK-STIK format)
Packaging Type Single
Species Escherichia coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.