Learn More
Description
Support formal quality assurance programs by serving as quality control challenges to verify performance of stain reagents, staining methods and the performance of personnel interpreting microscopic examinations.
Acid Fast Control SlideProvides two air-dried and methanol-fixed droplets within two etched circles
- Provides a single, air-dried and methanol-fixed fecal smear containing Acid Fast-Positive Cryptosporidium and acid fast-negative intestinal bacteria
- Provides a single, air-dried and methanol-fixed preparation containing Candida albicans, a derivative of ATCC™ 10231™†
- Provides a single, air dried, methanol-fixed, preparation containing Candida albicans, a derivative of ATCC™ 10231™† with leucocytes and erythrocytes
- Provides two air-dried and methanol-fixed droplets within two etched circles
- Circle nearest the label contains an acid-fast positive Mycobacterium gordonae, a derivative of ATCC™ 14470™†
- Circle farthest contains an acid fast-negative Erysipelothrix rhusiopathiae, a derivative of ATCC™ 19414™†
- Pneumocystis carinii Control Slide
- Provides a single, air-dried and methanol-fixed rat lung tissue preparation containing Pneumocystis
- SAF-Preserved Fecal Smear Control
- Provides an air-dried, SAF preserved, fecal smear containing a representative intestinal protozoa, usually Giardia lamblia
- S. aureus and E. coli Control Slide
- Provides two air-dried and methanol-fixed droplets within two etched circles
- Circle nearest the label contains a droplet of a Gram-Positive Staphylococcus aureus derivative of ATCC™ 25923™†
- Circle furthest from the label contains a droplet of a Gram-negative Escherichia coli derivative of ATCC™ 25922™†
- Provides an air-dried, Zinc PVA-preserved, fecal smear containing a representative intestinal protozoa, usually Giardia lamblia
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
Specifications
| Certifications/Compliance | CE |
| Format | Control Slide |
| Media Type | Slide |
| Product Type | Control Slide |
| For Use With (Application) | Microscopic examination |
| Quantity | 10 Slides |
| Specimen Type | Zinc PVA-Preserved Fecal Smear |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
