Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Microbiology QC Slides
SDP

Catalog No. 23023229 Shop All Microbiologics Products
Change view
Click to view available options
Specimen Type:
Acid Fast
Blood Parasite
Cryptosporidium
FYC Control Slide (Culture Isolates)
MYC-D (Clinical Samples)
Pneumocystis
S. aureis 25923 and E. coli 25922
SAF-Preserved Fecal Smear
Zinc PVA-Preserved Fecal Smear
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Specimen Type
23-023229 SAF-Preserved Fecal Smear
23-023223 S. aureis 25923 and E. coli 25922
23-023228 Zinc PVA-Preserved Fecal Smear
23-023221 Acid Fast
23-023227 Cryptosporidium
23-023226 FYC Control Slide (Culture Isolates)
23-023225 MYC-D (Clinical Samples)
23-023233 Pneumocystis
16-230-422 Blood Parasite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 23-023229 Supplier Microbiologics™ Supplier No. SL1510
Add to Cart
Add to Cart

Microscope slide preparations with one or two droplets of an air-dried and methanol-fixed organism, or a single smear containing a designated organism population

Support formal quality assurance programs by serving as quality control challenges to verify performance of stain reagents, staining methods and the performance of personnel interpreting microscopic examinations.

Acid Fast Control Slide

  • Provides two air-dried and methanol-fixed droplets within two etched circles
  • Circle nearest the label contains a droplet of an acid fast-positive Mycobacterium gordonae, a derivative of ATCC™ 14470™†
  • Circle furthest from the label contains a droplet of an acid fast-positive Cryptosporidium in a fecal sample and also contains acid fast-negative intestinal bacteria
Blood Parasite Control Slide
  • Provides an air-dried, methanol-fixed, blood parasite smear containing Plasmodium, Babesia or Trypanosoma
  • Cryptosporidium Control Slide
    • Provides a single, air-dried and methanol-fixed fecal smear containing Acid Fast-Positive Cryptosporidium and acid fast-negative intestinal bacteria
  • FYC Control Slide
    • Provides a single, air-dried and methanol-fixed preparation containing Candida albicans, a derivative of ATCC™ 10231™†
  • MYC-D Control Slide
    • Provides a single, air dried, methanol-fixed, preparation containing Candida albicans, a derivative of ATCC™ 10231™† with leucocytes and erythrocytes
  • Mycobacterium Control Slide
    • Provides two air-dried and methanol-fixed droplets within two etched circles
    • Circle nearest the label contains an acid-fast positive Mycobacterium gordonae, a derivative of ATCC™ 14470™†
    • Circle farthest contains an acid fast-negative Erysipelothrix rhusiopathiae, a derivative of ATCC™ 19414™†
    • Pneumocystis carinii Control Slide
      • Provides a single, air-dried and methanol-fixed rat lung tissue preparation containing Pneumocystis
    • SAF-Preserved Fecal Smear Control
      • Provides an air-dried, SAF preserved, fecal smear containing a representative intestinal protozoa, usually Giardia lamblia
    • S. aureus and E. coli Control Slide
      • Provides two air-dried and methanol-fixed droplets within two etched circles
      • Circle nearest the label contains a droplet of a Gram-Positive Staphylococcus aureus derivative of ATCC™ 25923™†
      • Circle furthest from the label contains a droplet of a Gram-negative Escherichia coli derivative of ATCC™ 25922™†
      Zinc PVA-Preserved Fecal Smear Control
      • Provides an air-dried, Zinc PVA-preserved, fecal smear containing a representative intestinal protozoa, usually Giardia lamblia

    †The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

    Specifications

    Certifications/Compliance CE
    Format Control Slide
    Media Type Slide
    Product Type Control Slide
    For Use With (Application) Microscopic examination
    Quantity 10 Slides
    Specimen Type SAF-Preserved Fecal Smear
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.