Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Proteus mirabilis ATCC™ 21100™
SDP

Catalog No. 23017205 Shop All Microbiologics Products
Change view
Click to view available options
Packaging Type:
KWIK STIK
LYFO DISK
Product Type:
Quality Control Organism
Quantity:
2/Pk.
6/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type Product Type Quantity
23-017-205 LYFO DISK Quality Control Organism 6/Pk.
23-017-206 KWIK STIK Quality Control Organism 2/Pk.
23-017-204 KWIK STIK Quality Control Organism 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-017-205 Supplier Microbiologics™ Supplier No. 01116L
This item is not returnable. View return policy

For culture purposes and QC applications

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features single strain of Proteus mirabilis in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
  • For added convenience, peel-off identification label is included for easy documentation
  • Available in pack of 2 or 6
LYFO DISK™
  • Reference stock culture option; lyophilized pellet containing single strain of Proteus mirabilis
  • Easy to use; re-hydrate pellet and inoculate
  • Available in re-sealable vial containing 6 pellets
  • ATCC™ 21100™

Specifications

ATCC® ATCC 21100
Biosafety Level 2
Format LYFO-DISK
Product Type Quality Control Organism
Packaging Type LYFO DISK
Quantity 6/Pk.
Species Proteus mirabilis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.