Learn More
Microbiologics™ RapID ONE QC Set
Catalog No. 23001671 Shop All Fisherbrand Products
Change view
Click to view available options
KWIK-STIK; 2/Pk.
Catalog No.
23-001-671 KWIK-STIK; 2/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-671
This product is either new or inactive, and cannot be ordered online.
This item is not returnable. View return policy
Includes
Enterobacter aerogenes ATCC™ 13048™†, Escherichia coli ATCC™ 25922™†, Pseudomonas aeruginosa ATCC™ 27853™†, Proteus vulgaris ATCC™ 6380™†
Microorganism sets for culture purposes and QC applications
  • Commercial sets contain the manufacturer recommended quality control strains for bacterial identification systems
  • Ordering microorganisms in sets simplifies the ordering process
  • Two convenient packaging options
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.