Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 13311 EZ-Accu Shot™ Kit†
SDP

Catalog No. 23201361 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Salmonella enterica subsp. enterica serovar typhimurium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-201-361 Salmonella enterica subsp. enterica serovar typhimurium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-201-361 Supplier Microbiologics™ Supplier No. 0421A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Five pellets of a single enumerated strain, five vials of hydrating fluid (1.2mL per vial) and a peel-off Certificate of Assay

For performing Growth Promotion Tests of culture media with ease

EZ-Accu Shot™

  • Quantitative microorganism preparation
  • Lyophilized microorganism preparation
  • Biosafety level: 1
  • No dilution required
  • Up to eight hours of stability
  • Adds efficiency and flexibility to QC laboratorie's busy schedules
  • Deliver 10 to 100 CFU per 0.1mL of hydrated suspension as recommended by the USP/JP/Ph. Eur
  • ATCC™ 13311™†

Specifications

ATCC® 13311
Biosafety Level 2
Format EZ-ACCU
For Use With (Application) For Growth Promotion testing.
Includes Five pellets of single enumerated strain; five vials of hydrating fluid (1.2 mL per vial); Peel-off certificate of assay
Species Salmonella enterica subsp. enterica serovar typhimurium

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.