Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features single strain of Staphylococcus aureus in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
- For added convenience, peel-off identification label is included for easy documentation
- Available in pack of 2 or 6
- Reference stock culture option; lyophilized pellet containing single strain of Staphylococcus aureus
- Easy to use; re-hydrate pellet and inoculate
- Available in re-sealable vial containing 6 pellets
- NCTC 13406
Specifications
Specifications
| ATCC® | ATCC BAA-2312 |
| Biosafety Level | 2 |
| Product Type | Quality Control Organism |
| Packaging Type | KWIK STIK |
| Quantity | 6/Pk. |
| Species | Staphylococcus aureus |