Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ ACCU SHOT
SDP

Catalog No. 16230502 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5 Pellts
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-230-502 5 Pellts
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-230-502 Supplier Microbiologics™ Supplier No. 0371A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For culture purposes and QC applications

  • Delivers 10-100 CFU per 0.1mL as required by USP/Ph. Eur./JP
  • Instant dissolve pellet reduces preparation time
  • No dilutions required for simplified test procedure
  • Up to 8 hours stability allows for ultimate flexibility
  • Refrigerated storage is easy and economical
  • Online Certificate of Analysis provides detailed strain information
  • Authentic, traceable strains at four passages (or fewer) from reference culture – meets USP/Ph. Eur./JP requirements
  • Technical Support experts available for guidance
  • Strains are less than or equal to four passages from reference culture

Specifications

ATCC® ATCC 12228™
Biosafety Level 1
Certifications/Compliance QC, online Certificate of Analysis
Concentration 10 to 100CFU per 0.1mL
Content And Storage Stored as directed
Form Lyophilized Pellet
For Use With (Equipment) Sterile Pipettes
Quantity 5 Pellts
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.