Learn More
SPEX CERTIPREP LLC 3P MICROBIOLOGY QC SAMPLE REFRIGE
Supplier:  SPEX CERTIPREP LLC 3P MICQC2
Encompass_Preferred
(NSI by Zeptometrix) Wastewater Microbiology QC Sample for Coliforms/E.coli, Ships Overnight Monday - Thursday, Must store in freezer
Catalog No. NC0227537