Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Scientific™ Microcentrifuge Tubes

Catalog No. 1184261
Change view
Click to view available options
Color:
Blue
Green
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11-842-61 Blue
11-842-23 Green
11-842-22 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-842-61 Supplier National Scientific™ Supplier No. CB170S GT

missing translation for 'validMainframeMsg'

Specifications

Certifications/Compliance RNase, DNase, Endotoxin-free
Autoclavable Autoclavable
Capacity (Metric) 1.7 mL
Color Blue
For Use With (Application) Ideal for custom gene libraries, protein work, cDNA synthesis, and nucleic acid amplification
Graduated Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.