Learn More
Cytiva (Formerly Pall Lab) Microcheck Beverage Monitor with Metricel Black membrane, 100 mL, 0.8 m, Black, Gridded, Sterile, Bulk Pack (50/box)
Catalog No. 502063091 Shop All Cytiva Products
Encompass
Easy-to-use Disposable Filter Funnel to Meet Microbial Analysis Needs for Beverages-Can convert to a Petri dish for culturing or easily remove the membrane for placing on separate agar dish.-Ready to use; simply remove from box and place directly on a filter manifold. No need for rubber stoppers or adapters.-Volume graduations are marked clearly for easy reading from any point of view.-Volume graduations marked externally on the funnel to prevent potential hold-up of organisms. External marks meet guidelines listed in Standard Methods for the Analysis of Water and Wastewater, current edition. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3091 Supplier Cytiva Supplier No. 4764
Easy-to-use Disposable Filter Funnel to Meet Microbial Analysis Needs for Beverages-Can convert to a Petri dish for culturing or easily remove the membrane for placing on separate agar dish.-Ready to use; simply remove from box and place directly on a filter manifold. No need for rubber stoppers or adapters.-Volume graduations are marked clearly for easy reading from any point of view.-Volume graduations marked externally on the funnel to prevent potential hold-up of organisms. External marks meet guidelines listed in Standard Methods for the Analysis of Water and Wastewater, current edition. This product is not approved for use in medical, clinical, surgical or other patient applications.