Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Life Sciences™ MicroCheck II Beverage Monitor

Catalog No. 09927603 Shop All Cytiva Products
Change view
Click to view available options
Capacity (Metric):
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-927-603 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-927-603 Supplier Cytiva Life Sciences™ Supplier No. 4761ME
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easy to use, versatile, disposable filter funnels for microbial analysis.

Can convert to a Petri dish for culturing or easily remove the membrane for placing on separate agar dish.

  • Can convert to a Petri dish for culturing or easily remove the membrane for placing on separate agar dish.
  • Ready to use: simply remove from box and place directly on a filter manifold.
  • No need for rubber stoppers or adapters.
  • Volume graduations are marked clearly for easy reading from any point of view.
  • Volume graduations marked externally on the funnel to prevent potential hold-up of organisms.
  • External marks meet guidelines listed in Standard Methods for the Analysis of Water and Wastewater, current edition.
  • Filter Media: GN Metricel™ (mixed cellulose esters) and Metricel™ Black (modified polyethersulfone) membranes
  • Cylinder and Base: Polypropylene
  • Cover: Polystyrene
  • Plug: Polyethylene
  • Funnel Adapter: Polyethylene
  • Support Pad: Cellulose
  • Height: 7.6 cm (3.0 in.)
  • Diameter: 6.1 cm (2.4 in.)
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 100 mL
Description Beverage monitor
Quantity 50 Funnels
Product Type Filtering Funnel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.