Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 2000 Shoe Covers - Model 417

Catalog No. 18167007 Shop All Ansell Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-167-007 Large
18-167-008 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-167-007 Supplier Ansell Supplier No. WH20B9241700
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Breathable microporous laminate technology provides protection from low hazard liquid spray and fine particulates.

  • Proven protective barrier to low concentration liquid chemicals, liquid & particulate biological hazards
  • Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
TRUSTED_SUSTAINABILITY

Specifications

Size Large
Material Microporous Polyethylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type ESD PVC
Model 417
Seam Type Bound
Style AlphaTec™ 2000
For Use With (Application) Life Sciences, Metal Fabrication, Food Processing
Certifications/Compliance (Europe) EN ISO 13688:2013
Product Type Shoe Covers
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.