Learn More
Description
- Protective multi-layer barrier fabric effective against numerous chemicals
- Highly visible bright yellow color for improved worker safety
- Lightweight and durable
- Applications:
- Unexpected leakages spills or other releases
- Loading and unloading trucks and vehicles
- Product inspection
- Cleaning of plant and machines
Specifications
Specifications
| Type | Protective Apron |
| Size | Universal |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Closure Type | Ties |
| Certifications/Compliance | CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126) |
| Dimensions (L x W) | 26 x 31 in. |
| Style | Bib Apron |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.