Learn More
Ansell AlphaTec™ 2000 Standard Lab Coats, Bound
Catalog No. 19500912 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-912 5X-Large
19-500-905 Small
19-500-906 Medium
19-500-907 Large
19-500-908 X-Large
19-500-909 2X-Large
19-500-910 3X-Large
19-500-911 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-500-912 Supplier Ansell Supplier No. WH20B9220909
Add to Cart
Add to Cart
This item is not returnable. View return policy
Low-linting lab coats provide a protective barrier to low concentration liquid chemicals and liquid and particulate biological hazards.
  • Protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
  • Free of natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Resistant Lab Coat
Size 5X-Large
Gender Unisex
Material Microporous Polyethylene Laminate
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
Style Standard Length
No. of Pockets 2
Latex Free Yes
Model 209
Seam Type Bound
Certifications/Compliance ASTM F1670, ASTM F1671
Certifications/Compliance (Europe) CE CATEGORY III (Type PB (6)-B, EN 14126)
For Use With (Application) Food Processing, Metal Fabrication, Cleanroom Cleaning and Preparing, Production Line Support and Maintenance, Veterinary
Packaging Type Individually Packaged
Autoclavable Not Autoclavable
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.