Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ MICROCHEM™ by AlphaTec™ Overboots

Catalog No. 19010349 Shop All Ansell Products
Change view
Click to view available options
Size:
42 to 46
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-349 42 to 46
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-010-349 Supplier Ansell™ Supplier No. WH20B9240700
  • $1,788.38
    $1,056.65 / Case of 150

    Save $731.73

    41% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ansell™ MICROCHEM™ by AlphaTec™ Overboots are designed with superior breathable microporous laminate technology.

  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Bound seams for improved overall protection
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
  • Free of natural rubber latex
  • Applications:
    • Clean room cleaning and preparing
    • Production line support and maintenance
    • Maintenance of plant and machines
    • Veterinary Services
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY I
Closure Type Tie fastening; Elastic on top of boot
Color White
Material Polyethylene
Seam Type Bound
Size 42 to 46
Weight (English) 63 gsm
Packaging Quantity 300/Cs.
Resistant To Liquid, Particles
Latex Free Yes
Ankle Style Elastic
Product Type Overboot
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.