Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROCHEM™ by AlphaTec™ Overboots

Catalog No. 19010347 Shop All Ansell Products
Change view
Click to view available options
Size:
12 to 14
8 to 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-347 8 to 12
19-010-348 12 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-010-347 Supplier Ansell Supplier No. WH15S9242200
This item is not returnable. View return policy

Ansell™ MICROCHEM™ by AlphaTec™ Overboots are air and water vapor permeable overboots with Sure Step sole and elastic foot opening

Air and water vapour permeable (breathable) to help reduce the risk of heat stress
TRUSTED_SUSTAINABILITY

Specifications

Size 8 to 12
Material SMS
Color White
Disposable Single-use
Closure Type Elastic Ankle
Seam Type Serged
For Use With (Application) Inspection, Asbestos Related Work, Powder Handling
Certifications/Compliance (Europe) EN 1073-2:2002; Type 5 EN ISO 13982-1:2004 + A1:2010; Type 6 EN 13034:2005 + A1:2009
Packaging Quantity 200/Cs.
Weight (Metric) 48 gsm
Product Type Boot Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.