Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ MICROCHEM™ by AlphaTec™ Overboots with Multi-Layer Chemical Barrier

Catalog No. 19010351 Shop All Ansell Products
Change view
Click to view available options
Size:
12 to 14
8 to 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-351 12 to 14
19-010-350 8 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-010-351 Supplier Ansell™ Supplier No. YE30W9240605
This item is not returnable. View return policy

Ansell™ MICROCHEM™ by AlphaTec™ Overboots provide an effective barrier against a range of inorganic chemicals and biological agents

  • Durable
  • Highly visible bright yellow color for improved worker safety
  • Lightweight and durable
  • Applications:
    • Unexpected leakages spills or other releases
    • Loading and unloading trucks and vehicles
    • Product inspection
    • Cleaning of plant and machines
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY I
Closure Type Elastic to top of boot with tie fastenings
Color Yellow
Material Multi-layer Non-woven Barrier Laminate
Seam Type Welded
Size 12 to 14
Packaging Quantity 200/case
Resistant To Liquid, Particles
Product Type Overboot

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.