Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ MICROCHEM™ by AlphaTec™ Oversleeves

Catalog No. 19500750 Shop All Ansell Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-500-750 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-500-750 Supplier Ansell™ Supplier No. YE30W92600

missing translation for 'validMainframeMsg'

Ansell™ MICROCHEM™ by AlphaTec™ Oversleeves feature comfortable multi-layer chemical barrier

  • Protective multi-layer barrier fabric effective against numerous chemicals
  • Highly visible bright yellow color for improved worker safety
  • Lightweight and durable
  • Applications:
    • Unexpected leakages spills or other releases
    • Loading and unloading trucks and vehicles
    • Product inspection
    • Cleaning of plant and machines
TRUSTED_SUSTAINABILITY

Specifications

Product Type Oversleeves
Color Yellow
Material Multi-layer Non-woven Barrier Laminate Fabric
Certifications/Compliance CE CATEGORY I
Size Universal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.