Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Colder Products Company MicroCNX™ Connector

Catalog No. 05113180 Shop All Colder Products Company Products
Change view
Click to view available options
Hose Barb:
1/16 in.
1/8 in.
3/32 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hose Barb
05-113-180 1/16 in.
05-113-181 1/8 in.
05-113-182 3/32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 05-113-180 Supplier Colder Products Company Supplier No. CNX17001HT
This item is not returnable. View return policy

The MicroCNX™ line introduces a new category of aseptic micro-connectors that provide a simple, efficient method of connecting tubing for small format biomanufacturing assemblies.

  • The MicroCNX line is engineered specifically for the challenging conditions of biologic media transfer in bioprocessing, cell therapy and gene therapy applications.
  • The Pinch-Click-Pull connection process reduces risk of operator error.
  • Eases single-use systems specifications with one part number for both halves.
  • CPC click allows for audible confirmation of assembly with no additional hardware required.

Specifications

Certifications/Compliance USP Class VI, ADCF
Material Polycarbonate
Min. Operating Temperature (English) 34°F
Min. Operating Temperature (Metric) 1°C
Hose Barb 1/16 in.
Sterilization Method Gamma, Autoclave
Height (Metric) 23.6 mm
Height (English) 0.93 in.
Material (Seal) Platinum-cured Silicone
Color White, Teal
Max. Operating Temperature (English) 104°F
Max. Operating Temperature (Metric) 40°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.