Learn More
MilliporeSigma™ Microcon™ Biomax™ MWCO Variety Pack
Catalog No. MPEVAR024 Shop All MilliporeSigma Products
Change view
Click to view available options
MWCO:
5, 10, 30, 50, 100, 300 kDa
Catalog No. MWCO
MPEVAR024 5, 10, 30, 50, 100, 300 kDa
1 options
Catalog No. MPEVAR024 Supplier MilliporeSigma™ Supplier No. MPEVAR024
Only null left
Add to Cart
Add to Cart
These Microcon™ Centrifugal Filter with Biomax™ Membrane are 0.5 mL centrifugal filters that are ideal for concentration and desalting of biomolecules.
  • Filter Type: Ultrafiltration
  • Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes
  • High recovery for small volumes with reverse spin (concentration factor less than 20X)
  • Convenient storage of filtrate or concentrated sample in standard microfuge tube
  • Concentration factors up to 100X
  • Biomax™ membrane
  • Fast processing
TRUSTED_SUSTAINABILITY
Specifications
Filter Type Ultrafiltration
Material (Membrane) PES
MWCO 5, 10, 30, 50, 100, 300 kDa
Sterility Non-sterile
For Use With (Application) These 0.5 mL centrifugal filters enable the concentration of biomolecules with a variety of membrane NMWL′s. Microcon centrifugal filters are ideal for concentration and desalting.
Includes 10 kDa NMWCO, 100 kDa NMWCO, 30 kDa NMWCO, 300 kDa NMWCO, 5 kDa NMWCO, 50 kDa NMWCO