Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microcrystalline Cellulose, NF, BP, JP, Spectrum™ Chemical
SDP

Catalog No. 18604600 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-600 50 kg Poly Bottle
18-604-596 100 g Poly Bottle
18-604-597 500 g Poly Bottle
18-604-598 2.5 kg Poly Bottle
18-604-599 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-600 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C457850KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

C4578, 9004-34-6

Spectrum™ Chemical Microcrystalline Cellulose, NF, BP, JP

Chemical Identifiers

CAS 9004-34-6
Molecular Formula C14H26O11
Molecular Weight (g/mol) 370.35
InChI Key PTHCMJGKKRQCBF-UHFFFAOYNA-N
IUPAC Name 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
SMILES COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O

Specifications

CAS Max % 1
Quantity 50 kg
Packaging Poly Bottle
Grade Multi-Compendial
Chemical Name or Material Microcrystalline Cellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.