Learn More
Enzo Life Sciences Microcystin-LR, (100µg), CAS Number: 101043-37-2
SDP
Supplier:  Enzo Life Sciences ALX350012C100
Encompass_Preferred
Inhibitor of PP1 and PP2A. Heptapeptide ester hepatotoxin. Tumor promoter. Equally potent and selective inhibitor of protein phosphatase 1 (PP1) and 2A (PP2A). PP2B is less sensitive and PP2C is not inhibited up to 4µM. Useful for affinity-purification of PP2A. The product is not cell permeable except in liver cells, which appear to have a functional uptake system. Is absorbed by hepatocytes.7.7. Long Term Storage: -20°C
Catalog No. NC0198080
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.