Learn More
Enzo Life Sciences Microcystin-YR (25µg). CAS: 101064-48-6
SDP
Supplier:  Enzo Life Sciences ALX350044C025
Encompass_Preferred
Analog of microcystin-LR (ALX-350-012) with Tyr substituted in place of Leu. As for all microcystins the conjugated double bonds in the ADDA moiety cause a characteristic absorption maximum at 238nm. The Tyr residue in position 2 of microcystin-YR confers an absorption maximum at 232nm. Potent inhibitor of eukaryotic protein phosphatases 1 and 2A. Useful as a reference compound in environmental analysis. The hydroxyl group of the Tyr residue may prove useful for linking microcystin-YR via conjugation to other chemicals. Inhibits the synthesis of proteases such as cathepsin D and L, and arginine aminopeptidase. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8707
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.